The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145819.1 | NP_001139291.1 | 11 - 12 | 1502 | 122 |
NM_033326.3 | NP_201583.2 | 11 - 12 | 1508 | 122 | |
GenBank mRNA | AF309034.1 | - | 11 - 12 | 1491 | 122 |
BC047064.1 | - | 11 - 12 | 1498 | 122 |