Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SOX6
Entrez Gene ID
55553
Gene Name
SRY-box 6
Gene Aliases
HSSOX6, SOXD
Chromosome Location
Chr.11: 15966449 - 16476388 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145811.1 NP_001139283.1 13 - 14 2077 92
NM_001145819.1 NP_001139291.1 14 - 15 2039 92
NM_017508.2 NP_059978.1 13 - 14 2099 92
NM_033326.3 NP_201583.2 14 - 15 1985 92
GenBank mRNA AF309034.1 - 14 - 15 1968 92
AL136780.1 - 13 - 14 2078 92
BC047064.1 - 14 - 15 2035 92
X65663.1 - 1 - 2 74 92

View Full Product Details