Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIMCH1
Entrez Gene ID
22998
Gene Name
LIM and calponin homology domains 1
Gene Aliases
LIMCH1A, LMO7B
Chromosome Location
Chr.4: 41359607 - 41700044 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112717.2 NP_001106188.1 15 - 16 2198 60
NM_001112718.2 NP_001106189.1 15 - 16 2198 60
NM_001112719.2 NP_001106190.1 10 - 11 1983 60
NM_001112720.2 NP_001106191.1 9 - 10 1947 60
NM_001289122.1 NP_001276051.1 14 - 15 2162 60
NM_001289124.1 NP_001276053.1 14 - 15 2207 60
NM_014988.3 NP_055803.2 15 - 16 2198 60
XM_005248057.1 XP_005248114.1 22 - 23 3830 60
XM_005248058.1 XP_005248115.1 22 - 23 3830 60
XM_005248060.1 XP_005248117.1 22 - 23 3830 60
XM_005248061.1 XP_005248118.1 22 - 23 3830 60
XM_005248067.2 XP_005248124.1 20 - 21 4704 60
XM_005248072.1 XP_005248129.1 15 - 16 2198 60
XM_005248074.1 XP_005248131.1 14 - 15 2162 60
XM_005248075.1 XP_005248132.1 14 - 15 2162 60
XM_006713996.1 XP_006714059.1 22 - 23 3830 60
XM_011513642.1 XP_011511944.1 22 - 23 3830 60
XM_011513643.1 XP_011511945.1 22 - 23 3830 60
XM_011513644.1 XP_011511946.1 22 - 23 3830 60
XM_011513645.1 XP_011511947.1 22 - 23 3830 60
XM_011513646.1 XP_011511948.1 22 - 23 3830 60
XM_011513648.1 XP_011511950.1 21 - 22 3587 60
XM_011513649.1 XP_011511951.1 17 - 18 3489 60
XM_011513651.2 XP_011511953.1 20 - 21 4709 60
XM_011513653.1 XP_011511955.1 15 - 16 2198 60
XM_011513654.1 XP_011511956.1 14 - 15 2162 60
XM_011513655.1 XP_011511957.1 9 - 10 1821 60
XM_011513656.1 XP_011511958.1 10 - 11 1857 60
XM_011513657.1 XP_011511959.1 9 - 10 1821 60
XM_011513658.2 XP_011511960.1 14 - 15 2423 60
XM_017007894.1 XP_016863383.1 22 - 23 3830 60
XM_017007895.1 XP_016863384.1 22 - 23 3830 60
XM_017007896.1 XP_016863385.1 22 - 23 3830 60
XM_017007897.1 XP_016863386.1 22 - 23 3830 60
XM_017007898.1 XP_016863387.1 15 - 16 2198 60
XM_017007899.1 XP_016863388.1 15 - 16 2198 60
XM_017007900.1 XP_016863389.1 15 - 16 2198 60
XM_017007901.1 XP_016863390.1 15 - 16 2198 60
XM_017007902.1 XP_016863391.1 15 - 16 2198 60
XM_017007903.1 XP_016863392.1 10 - 11 1857 60
XM_017007904.1 XP_016863393.1 10 - 11 1857 60
XM_017007905.1 XP_016863394.1 10 - 11 1857 60
GenBank mRNA AB029025.2 - 15 - 16 2196 60
AK294774.1 - 9 - 10 2010 60
AK295784.1 - 8 - 9 1536 60
AK295836.1 - 9 - 10 1646 60
AK298915.1 - 10 - 11 1853 60
AK299297.1 - 9 - 10 1817 60
AK299835.1 - 9 - 10 1681 60
AK302674.1 - 2 - 3 449 60
AK309846.1 - 9 - 10 1889 60
AL831962.2 - 20 - 21 3585 60
BC023546.2 - 12 - 13 2051 60
BX537916.1 - 9 - 10 1817 60
BX640692.1 - 8 - 9 1575 60
CR749205.1 - 10 - 11 1853 60
CR933645.1 - 15 - 16 2199 60
CR936601.1 - 14 - 15 2133 60
CR936661.1 - 14 - 15 2102 60
CR936664.1 - 15 - 16 2199 60

View Full Product Details