Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIMCH1
Entrez Gene ID
22998
Gene Name
LIM and calponin homology domains 1
Gene Aliases
LIMCH1A, LMO7B
Chromosome Location
Chr.4: 41359607 - 41700044 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112717.2 NP_001106188.1 8 - 9 997 101
NM_001112718.2 NP_001106189.1 8 - 9 997 101
NM_001112719.2 NP_001106190.1 3 - 4 782 101
NM_014988.3 NP_055803.2 8 - 9 997 101
XM_005248072.1 XP_005248129.1 8 - 9 997 101
XM_011513653.1 XP_011511955.1 8 - 9 997 101
XM_011513656.1 XP_011511958.1 3 - 4 656 101
XM_017007898.1 XP_016863387.1 8 - 9 997 101
XM_017007899.1 XP_016863388.1 8 - 9 997 101
XM_017007900.1 XP_016863389.1 8 - 9 997 101
XM_017007901.1 XP_016863390.1 8 - 9 997 101
XM_017007902.1 XP_016863391.1 8 - 9 997 101
XM_017007903.1 XP_016863392.1 3 - 4 656 101
XM_017007904.1 XP_016863393.1 3 - 4 656 101
XM_017007905.1 XP_016863394.1 3 - 4 656 101
GenBank mRNA AB029025.2 - 8 - 9 995 101
AK295836.1 - 2 - 3 445 101
BC023546.2 - 5 - 6 850 101
CR749205.1 - 3 - 4 652 101
CR933645.1 - 8 - 9 998 101
CR936658.1 - 8 - 9 976 101
CR936664.1 - 8 - 9 998 101

View Full Product Details