The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143996.1 | NP_001137468.1 | 6 - 7 | 867 | 84 |
NM_001143997.1 | NP_001137469.1 | 5 - 6 | 662 | 84 | |
NM_018245.2 | NP_060715.2 | 7 - 8 | 1038 | 84 | |
XM_011539946.1 | XP_011538248.1 | 7 - 8 | 923 | 84 | |
XM_011539947.1 | XP_011538249.1 | 7 - 8 | 1129 | 84 | |
XM_017016401.1 | XP_016871890.1 | 7 - 8 | 1313 | 84 | |
GenBank mRNA | AB033116.3 | - | 6 - 7 | 901 | 84 |
AK001713.1 | - | 7 - 8 | 982 | 84 | |
AK225558.1 | - | 7 - 8 | 969 | 84 | |
AK290226.1 | - | 6 - 7 | 1011 | 84 | |
AK296551.1 | - | 6 - 7 | 776 | 84 | |
AK303729.1 | - | 5 - 6 | 609 | 84 | |
AK307759.1 | - | 6 - 7 | 797 | 84 | |
BC026320.1 | - | 7 - 8 | 957 | 84 | |
DA411851.1 | - | 3 - 4 | 430 | 84 | |
DA416844.1 | - | 4 - 5 | 448 | 84 |