The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143996.1 | NP_001137468.1 | 9 - 10 | 1267 | 85 |
NM_001143997.1 | NP_001137469.1 | 8 - 9 | 1062 | 85 | |
NM_018245.2 | NP_060715.2 | 10 - 11 | 1438 | 85 | |
XM_011539946.1 | XP_011538248.1 | 10 - 11 | 1323 | 85 | |
XM_011539947.1 | XP_011538249.1 | 10 - 11 | 1529 | 85 | |
XM_017016401.1 | XP_016871890.1 | 10 - 11 | 1713 | 85 | |
XM_017016402.1 | XP_016871891.1 | 10 - 11 | 1340 | 85 | |
XM_017016403.1 | XP_016871892.1 | 8 - 9 | 1086 | 85 | |
GenBank mRNA | AB033116.3 | - | 9 - 10 | 1301 | 85 |
AK001713.1 | - | 10 - 11 | 1382 | 85 | |
AK290226.1 | - | 9 - 10 | 1411 | 85 | |
AK296551.1 | - | 9 - 10 | 1176 | 85 | |
AK303729.1 | - | 8 - 9 | 1009 | 85 | |
AK307759.1 | - | 9 - 10 | 1197 | 85 | |
BC026320.1 | - | 10 - 11 | 1357 | 85 |