The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000923.4 | NP_000914.2 | 5 - 6 | 784 | 65 |
NM_001098818.2 | NP_001092288.1 | 4 - 5 | 509 | 65 | |
NM_001098819.2 | NP_001092289.1 | 4 - 5 | 562 | 65 | |
XM_011528056.2 | XP_011526358.1 | 4 - 5 | 515 | 65 | |
XM_011528058.2 | XP_011526360.1 | 2 - 3 | 91 | 65 | |
XM_017026866.1 | XP_016882355.1 | 5 - 6 | 793 | 65 | |
GenBank mRNA | AK295050.1 | - | 4 - 5 | 1025 | 65 |
AK300983.1 | - | 4 - 5 | 472 | 65 | |
AY429556.1 | - | 1 - 2 | 31 | 65 | |
BC109068.2 | - | 1 - 2 | 57 | 65 | |
U66346.1 | - | 5 - 6 | 784 | 65 | |
U88712.1 | - | 4 - 5 | 562 | 65 | |
U88713.1 | - | 4 - 5 | 509 | 65 | |
Z46632.1 | - | 4 - 5 | 656 | 65 |