Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZCCHC10
Entrez Gene ID
54819
Gene Name
zinc finger CCHC-type containing 10
Gene Aliases
-
Chromosome Location
Chr.5: 132996985 - 133026621 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300816.2 NP_001287745.1 2 - 3 168 104
NM_001300817.2 NP_001287746.1 2 - 3 168 104
NM_001300818.2 NP_001287747.1 2 - 3 168 104
NM_001300822.2 NP_001287751.1 2 - 3 168 104
GenBank mRNA AK122970.1 - 2 - 3 113 104
BC022443.1 - 2 - 3 109 104
BU165290.1 - 2 - 3 99 104
CB112418.1 - 2 - 3 109 104

View Full Product Details