Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MYEF2
- Entrez Gene ID
-
50804
- Gene Name
- myelin expression factor 2
- Gene Aliases
-
HsT18564, MEF-2, MST156, MSTP156, myEF-2
- Chromosome Location
-
Chr.15: 48134632 - 48178517 on Build GRCh38
View Full Product Details