Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GSE1
Entrez Gene ID
23199
Gene Name
Gse1 coiled-coil protein
Gene Aliases
CRHSP24, KIAA0182
Chromosome Location
Chr.16: 85169512 - 85676206 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278184.1 NP_001265113.1 1 - 2 170 80
GenBank mRNA D80004.2 - 1 - 2 54 80

View Full Product Details