Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MKL2
Entrez Gene ID
57496
Gene Name
MKL1/myocardin like 2
Gene Aliases
MRTF-B, MRTFB, NPD001
Chromosome Location
Chr.16: 14071076 - 14266779 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308142.1 NP_001295071.1 11 - 12 1388 117
NM_014048.4 NP_054767.3 11 - 12 1389 117
XM_005255452.3 XP_005255509.1 12 - 13 1543 117
XM_005255453.4 XP_005255510.1 11 - 12 2027 117
XM_005255455.3 XP_005255512.1 9 - 10 1373 117
XM_006720908.3 XP_006720971.1 12 - 13 2189 117
XM_006720909.3 XP_006720972.1 12 - 13 1518 117
XM_006720912.3 XP_006720975.1 9 - 10 3328 117
XM_006720913.2 XP_006720976.1 12 - 13 1543 117
XM_006720914.2 XP_006720977.1 12 - 13 1543 117
XM_011522568.2 XP_011520870.1 11 - 12 1358 117
XM_011522569.2 XP_011520871.1 9 - 10 1155 117
XM_017023501.1 XP_016878990.1 11 - 12 1354 117
XM_017023502.1 XP_016878991.1 11 - 12 1411 117
XM_017023503.1 XP_016878992.1 10 - 11 4556 117
XM_017023504.1 XP_016878993.1 9 - 10 1373 117
XM_017023505.1 XP_016878994.1 9 - 10 3328 117
GenBank mRNA AB033069.2 - 3 - 4 395 117
AK294765.1 - 11 - 12 1380 117
AY374297.1 - 11 - 12 1366 117
BC136260.1 - 9 - 10 1233 117
BC171750.1 - 9 - 10 1233 117

View Full Product Details