The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272049.1 | NP_001258978.1 | 6 - 7 | 747 | 75 |
NM_016292.2 | NP_057376.2 | 7 - 8 | 906 | 75 | |
XM_011522345.1 | XP_011520647.1 | 7 - 8 | 798 | 75 | |
XM_017022851.1 | XP_016878340.1 | 6 - 7 | 729 | 75 | |
GenBank mRNA | AB209805.1 | - | 7 - 8 | 816 | 75 |
AF043254.1 | - | 7 - 8 | 808 | 75 | |
AF154108.1 | - | 7 - 8 | 821 | 75 | |
AJ890085.1 | - | 2 - 3 | 920 | 75 | |
AK057620.1 | - | 2 - 3 | 920 | 75 | |
AK093344.1 | - | 7 - 8 | 859 | 75 | |
AK223049.1 | - | 7 - 8 | 826 | 75 | |
AK223076.1 | - | 7 - 8 | 826 | 75 | |
AK223205.1 | - | 7 - 8 | 826 | 75 | |
AK299127.1 | - | 6 - 7 | 663 | 75 | |
BC001455.2 | - | 7 - 8 | 804 | 75 | |
BC018950.2 | - | 7 - 8 | 824 | 75 | |
BC023585.2 | - | 7 - 8 | 828 | 75 | |
JX481354.1 | - | 6 - 7 | 734 | 75 | |
U12595.1 | - | 7 - 8 | 774 | 75 |