The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164104.1 | NP_001157576.1 | 11 - 12 | 1835 | 128 |
NM_001164105.1 | NP_001157577.1 | 11 - 12 | 1912 | 128 | |
NM_018995.2 | NP_061868.1 | 11 - 12 | 1835 | 128 | |
XM_005261923.3 | XP_005261980.1 | 11 - 12 | 2113 | 128 | |
XM_011530696.1 | XP_011528998.1 | 11 - 12 | 1835 | 128 | |
XM_011530697.1 | XP_011528999.1 | 11 - 12 | 2113 | 128 | |
XM_011530698.1 | XP_011529000.1 | 10 - 11 | 1694 | 128 | |
XM_011530699.1 | XP_011529001.1 | 11 - 12 | 1797 | 128 | |
XM_011530700.2 | XP_011529002.1 | 11 - 12 | 2180 | 128 | |
XM_011530701.1 | XP_011529003.1 | 11 - 12 | 1835 | 128 | |
XM_011530702.1 | XP_011529004.1 | 10 - 11 | 1608 | 128 | |
XM_011530703.1 | XP_011529005.1 | 4 - 5 | 683 | 128 | |
XM_011530704.1 | XP_011529006.1 | 11 - 12 | 1835 | 128 | |
XM_017028833.1 | XP_016884322.1 | 11 - 12 | 1835 | 128 | |
XM_017028834.1 | XP_016884323.1 | 11 - 12 | 1835 | 128 | |
XM_017028835.1 | XP_016884324.1 | 2 - 3 | 249 | 128 | |
XM_017028836.1 | XP_016884325.1 | 2 - 3 | 206 | 128 | |
XM_017028837.1 | XP_016884326.1 | 11 - 12 | 1835 | 128 | |
GenBank mRNA | AF285604.1 | - | 11 - 12 | 1779 | 128 |
AK092675.1 | - | 5 - 6 | 671 | 128 | |
AK302401.1 | - | 11 - 12 | 1912 | 128 | |
AK303017.1 | - | 6 - 7 | 1049 | 128 | |
AL133068.2 | - | 1 - 2 | 154 | 128 | |
BC140944.1 | - | 12 - 13 | 1836 | 128 | |
BC150137.1 | - | 11 - 12 | 1777 | 128 | |
BC152539.1 | - | 11 - 12 | 1777 | 128 | |
CR456466.1 | - | 11 - 12 | 1758 | 128 |