The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164104.1 | NP_001157576.1 | 17 - 18 | 2439 | 95 |
NM_001164105.1 | NP_001157577.1 | 17 - 18 | 2516 | 95 | |
NM_018995.2 | NP_061868.1 | 17 - 18 | 2439 | 95 | |
XM_005261923.3 | XP_005261980.1 | 17 - 18 | 2717 | 95 | |
XM_011530696.1 | XP_011528998.1 | 17 - 18 | 2439 | 95 | |
XM_011530697.1 | XP_011528999.1 | 17 - 18 | 2717 | 95 | |
XM_011530698.1 | XP_011529000.1 | 16 - 17 | 2298 | 95 | |
XM_011530699.1 | XP_011529001.1 | 17 - 18 | 2401 | 95 | |
XM_011530700.2 | XP_011529002.1 | 17 - 18 | 2784 | 95 | |
XM_011530701.1 | XP_011529003.1 | 17 - 18 | 2439 | 95 | |
XM_011530702.1 | XP_011529004.1 | 16 - 17 | 2212 | 95 | |
XM_011530703.1 | XP_011529005.1 | 10 - 11 | 1287 | 95 | |
XM_017028833.1 | XP_016884322.1 | 17 - 18 | 2439 | 95 | |
XM_017028835.1 | XP_016884324.1 | 8 - 9 | 853 | 95 | |
XM_017028836.1 | XP_016884325.1 | 8 - 9 | 810 | 95 | |
GenBank mRNA | AF285604.1 | - | 17 - 18 | 2383 | 95 |
AK092675.1 | - | 11 - 12 | 1275 | 95 | |
AK302401.1 | - | 17 - 18 | 2516 | 95 | |
AL133068.2 | - | 7 - 8 | 758 | 95 | |
BC140944.1 | - | 18 - 19 | 2440 | 95 | |
BC150137.1 | - | 17 - 18 | 2381 | 95 | |
BC152539.1 | - | 17 - 18 | 2381 | 95 | |
CR456466.1 | - | 17 - 18 | 2362 | 95 |