Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SELO
Entrez Gene ID
83642
Gene Name
selenoprotein O
Gene Aliases
-
Chromosome Location
Chr.22: 50200979 - 50217616 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC020510.1 - 1 - 2 79 67

View Full Product Details