The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145811.1 | NP_001139283.1 | 8 - 9 | 1169 | 84 |
NM_017508.2 | NP_059978.1 | 8 - 9 | 1191 | 84 | |
GenBank mRNA | AL136780.1 | - | 8 - 9 | 1170 | 84 |