The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145811.1 | NP_001139283.1 | 10 - 11 | 1535 | 145 |
NM_017508.2 | NP_059978.1 | 10 - 11 | 1557 | 145 | |
GenBank mRNA | AL136780.1 | - | 10 - 11 | 1536 | 145 |