The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_201648.2 | NP_964011.2 | 5 - 6 | 641 | 117 |
XM_017017087.1 | XP_016872576.1 | 5 - 6 | 704 | 117 | |
GenBank mRNA | AB013093.1 | - | 5 - 6 | 590 | 117 |
AB665285.1 | - | 5 - 6 | 619 | 117 |