The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016938.4 | NP_058634.4 | 9 - 10 | 1233 | 134 |
NR_037718.1 | 9 - 10 | 1233 | 134 | ||
GenBank mRNA | AB030655.1 | - | 9 - 10 | 1070 | 134 |
AB209121.1 | - | 5 - 6 | 4536 | 134 | |
AF109121.1 | - | 9 - 10 | 1061 | 134 | |
AF124486.1 | - | 8 - 9 | 974 | 134 | |
AJ132819.2 | - | 8 - 9 | 974 | 134 | |
AK000980.1 | - | 9 - 10 | 1070 | 134 | |
AK075453.1 | - | 9 - 10 | 1039 | 134 | |
AK095794.1 | - | 2 - 3 | 1540 | 134 | |
AK098186.1 | - | 5 - 6 | 1973 | 134 | |
AK292079.1 | - | 9 - 10 | 1078 | 134 | |
AY358899.1 | - | 9 - 10 | 1035 | 134 | |
BC010456.1 | - | 9 - 10 | 1089 | 134 | |
BC018871.2 | - | 9 - 10 | 1110 | 134 | |
CR541934.1 | - | 8 - 9 | 974 | 134 |