The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016938.4 | NP_058634.4 | 1 - 2 | 254 | 124 |
NR_037718.1 | 1 - 2 | 254 | 124 | ||
GenBank mRNA | AB209121.1 | - | 1 - 2 | 928 | 124 |
AK000980.1 | - | 1 - 2 | 91 | 124 | |
AK292079.1 | - | 1 - 2 | 99 | 124 | |
BC010456.1 | - | 1 - 2 | 110 | 124 | |
BC018871.2 | - | 1 - 2 | 131 | 124 | |
BP382163.1 | - | 1 - 2 | 164 | 124 | |
DB475347.1 | - | 1 - 2 | 189 | 124 |