The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005396.4 | NP_005387.2 | 7 - 8 | 718 | 80 |
NR_103727.1 | 7 - 8 | 718 | 80 | ||
XM_017016339.1 | XP_016871828.1 | 6 - 7 | 702 | 80 | |
GenBank mRNA | AK308511.1 | - | 7 - 8 | 1202 | 80 |
AK308521.1 | - | 7 - 8 | 1212 | 80 | |
BC005989.2 | - | 6 - 7 | 692 | 80 | |
CR456949.1 | - | 6 - 7 | 692 | 80 | |
M93284.1 | - | 6 - 7 | 692 | 80 |