Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGFG2
Entrez Gene ID
3268
Gene Name
ArfGAP with FG repeats 2
Gene Aliases
HRBL, RABR
Chromosome Location
Chr.7: 100539211 - 100568220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017012114.1 XP_016867603.1 3 - 4 553 100
GenBank mRNA BC009393.2 - 3 - 4 566 100

View Full Product Details