Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGFG2
Entrez Gene ID
3268
Gene Name
ArfGAP with FG repeats 2
Gene Aliases
HRBL, RABR
Chromosome Location
Chr.7: 100539211 - 100568220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006076.4 NP_006067.3 11 - 12 1522 57
XM_005250306.2 XP_005250363.1 12 - 13 1541 57
XM_017012112.1 XP_016867601.1 11 - 12 1463 57
XM_017012114.1 XP_016867603.1 10 - 11 1221 57
GenBank mRNA AF015042.1 - 11 - 12 1446 57
AK314954.1 - 11 - 12 1508 57
BC017329.2 - 11 - 12 1474 57

View Full Product Details