The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024712.3 | NP_078988.2 | 7 - 8 | 873 | 54 |
GenBank mRNA | AK023886.1 | - | 6 - 7 | 637 | 54 |
AK056923.1 | - | 6 - 7 | 836 | 54 | |
AK300976.1 | - | 7 - 8 | 887 | 54 | |
BC034410.1 | - | 4 - 5 | 485 | 54 |