Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELMO3
Entrez Gene ID
79767
Gene Name
engulfment and cell motility 3
Gene Aliases
CED-12, CED12, ELMO-3
Chromosome Location
Chr.16: 67199125 - 67204024 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024712.3 NP_078988.2 9 - 10 1004 69
GenBank mRNA AK023886.1 - 8 - 9 768 69
AK056923.1 - 8 - 9 967 69
AK300976.1 - 9 - 10 1018 69
BC034410.1 - 6 - 7 616 69

View Full Product Details