The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267722.1 | NP_001254651.1 | 2 - 3 | 267 | 103 |
NM_006947.3 | NP_008878.3 | 2 - 3 | 267 | 103 | |
XM_005265765.4 | XP_005265822.3 | 2 - 3 | 986 | 103 | |
GenBank mRNA | AF038851.1 | - | 2 - 3 | 239 | 103 |
AF069765.1 | - | 2 - 3 | 237 | 103 | |
AF077019.1 | - | 2 - 3 | 227 | 103 | |
AK225430.1 | - | 2 - 3 | 230 | 103 | |
AK307397.1 | - | 2 - 3 | 230 | 103 | |
BC017057.1 | - | 2 - 3 | 255 | 103 | |
BC032609.1 | - | 2 - 3 | 245 | 103 | |
BC040134.1 | - | 2 - 3 | 248 | 103 | |
BC046143.1 | - | 2 - 3 | 239 | 103 | |
BC065018.1 | - | 2 - 3 | 239 | 103 | |
BC105583.1 | - | 2 - 3 | 255 | 103 | |
BX537991.1 | - | 2 - 3 | 247 | 103 | |
DB487204.2 | - | 2 - 3 | 267 | 103 | |
EU794663.1 | - | 2 - 3 | 237 | 103 |