Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C4orf36
Entrez Gene ID
132989
Gene Name
chromosome 4 open reading frame 36
Gene Aliases
-
Chromosome Location
Chr.4: 86876205 - 86897256 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144645.3 NP_653246.1 1 - 2 233 83
GenBank mRNA BC016746.1 - 1 - 2 209 83

View Full Product Details