The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198994.2 | NP_945345.2 | 11 - 12 | 1892 | 100 |
GenBank mRNA | AF540969.1 | - | 11 - 12 | 1892 | 100 |