The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001254734.1 | NP_001241663.1 | 9 - 10 | 1400 | 63 |
NM_198994.2 | NP_945345.2 | 9 - 10 | 1400 | 63 | |
GenBank mRNA | AF540969.1 | - | 9 - 10 | 1400 | 63 |
AF540970.1 | - | 9 - 10 | 1400 | 63 |