The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001059.2 | NP_001050.1 | 2 - 3 | 868 | 85 |
GenBank mRNA | AY462099.1 | - | 2 - 3 | 727 | 85 |
BC095527.1 | - | 2 - 3 | 724 | 85 | |
BC121806.1 | - | 2 - 3 | 828 | 85 | |
M89473.1 | - | 2 - 3 | 870 | 85 |