Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RIBC2
Entrez Gene ID
26150
Gene Name
RIB43A domain with coiled-coils 2
Gene Aliases
C22orf11, TRIB
Chromosome Location
Chr.22: 45413691 - 45432422 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015653.4 NP_056468.3 1 - 2 323 128
XM_017028766.1 XP_016884255.1 1 - 2 245 128
GenBank mRNA AK098586.1 - 1 - 2 323 128
AK225722.1 - 1 - 2 304 128
BC003024.1 - 1 - 2 285 128
BC009904.2 - 1 - 2 285 128

View Full Product Details