The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015653.4 | NP_056468.3 | 5 - 6 | 1100 | 61 |
XM_005261524.4 | XP_005261581.1 | 5 - 6 | 925 | 61 | |
XM_011530126.2 | XP_011528428.1 | 3 - 4 | 628 | 61 | |
GenBank mRNA | AK098586.1 | - | 5 - 6 | 1100 | 61 |
AK225722.1 | - | 5 - 6 | 1081 | 61 | |
AL050075.1 | - | 1 - 2 | 208 | 61 | |
AL442116.1 | - | 5 - 6 | 1051 | 61 | |
BC003024.1 | - | 5 - 6 | 1062 | 61 | |
BC009904.2 | - | 5 - 6 | 1062 | 61 | |
CR456438.1 | - | 3 - 4 | 694 | 61 | |
CR457398.1 | - | 3 - 4 | 685 | 61 |