The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178088.1 | NP_001171559.1 | 7 - 8 | 1149 | 79 |
NM_003898.3 | NP_003889.1 | 8 - 9 | 1226 | 79 | |
XM_005267198.4 | XP_005267255.1 | 7 - 8 | 1157 | 79 | |
XM_006715592.3 | XP_006715655.1 | 8 - 9 | 1226 | 79 | |
XM_011536224.1 | XP_011534526.1 | 8 - 9 | 1226 | 79 | |
XM_011536225.1 | XP_011534527.1 | 8 - 9 | 1266 | 79 | |
XM_011536226.2 | XP_011534528.1 | 2 - 3 | 2130 | 79 | |
XM_011536227.2 | XP_011534529.1 | 8 - 9 | 1226 | 79 | |
XM_011536230.2 | XP_011534532.1 | 8 - 9 | 1226 | 79 | |
GenBank mRNA | AB002346.2 | - | 7 - 8 | 962 | 79 |
AF039945.1 | - | 7 - 8 | 962 | 79 | |
AF318616.1 | - | 8 - 9 | 1203 | 79 | |
AK296242.1 | - | 7 - 8 | 1174 | 79 | |
AY152396.1 | - | 8 - 9 | 1128 | 79 | |
BX648749.1 | - | 7 - 8 | 1056 | 79 |