The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000485.2 | NP_000476.1 | 2 - 3 | 216 | 113 |
NM_001030018.1 | NP_001025189.1 | 2 - 3 | 216 | 113 | |
GenBank mRNA | BC106894.2 | - | 3 - 4 | 113 | |
BC106895.2 | - | 3 - 4 | 113 | ||
BC107151.2 | - | 2 - 3 | 206 | 113 | |
BM423481.1 | - | - | 240 | 113 | |
BM550173.1 | - | 2 - 3 | 239 | 113 | |
CR749423.1 | - | 2 - 3 | 207 | 113 |