The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000485.2 | NP_000476.1 | 4 - 5 | 432 | 66 |
GenBank mRNA | BC106894.2 | - | 1 - 2 | 66 | |
BC106895.2 | - | 1 - 2 | 66 | ||
BC107151.2 | - | 4 - 5 | 422 | 66 | |
BM423481.1 | - | - | 456 | 66 | |
CR749423.1 | - | 4 - 5 | 423 | 66 |