The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006355.4 | NP_006346.1 | 5 - 6 | 1327 | 90 |
XM_005248799.4 | XP_005248856.1 | 5 - 6 | 1056 | 90 | |
XM_005248800.4 | XP_005248857.1 | 5 - 6 | 1197 | 90 | |
XM_017010175.1 | XP_016865664.1 | 5 - 6 | 1216 | 90 | |
GenBank mRNA | AB209906.1 | - | 3 - 4 | 3617 | 90 |
AK313248.1 | - | 5 - 6 | 1214 | 90 | |
BC026930.1 | - | 4 - 5 | 1173 | 90 | |
BX640949.1 | - | 5 - 6 | 1244 | 90 | |
U90547.1 | - | 3 - 4 | 759 | 90 |