The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006355.4 | NP_006346.1 | 7 - 8 | 1464 | 93 |
XM_005248799.4 | XP_005248856.1 | 7 - 8 | 1193 | 93 | |
XM_005248800.4 | XP_005248857.1 | 7 - 8 | 1334 | 93 | |
GenBank mRNA | AB209906.1 | - | 5 - 6 | 3754 | 93 |
AK313248.1 | - | 7 - 8 | 1351 | 93 | |
BC026930.1 | - | 6 - 7 | 1310 | 93 | |
U90547.1 | - | 5 - 6 | 896 | 93 |