Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KNDC1
Entrez Gene ID
85442
Gene Name
kinase non-catalytic C-lobe domain containing 1
Gene Aliases
C10orf23, RASGEF2, bB439H18.3
Chromosome Location
Chr.10: 133160096 - 133226412 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152643.6 NP_689856.6 10 - 11 1734 95
XM_017016858.1 XP_016872347.1 10 - 11 2102 95
XM_017016859.1 XP_016872348.1 10 - 11 2105 95
GenBank mRNA AB000781.1 - 9 - 10 1630 95
AB051555.1 - 9 - 10 1828 95
AK090457.1 - 9 - 10 2641 95

View Full Product Details