The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152643.6 | NP_689856.6 | 4 - 5 | 511 | 112 |
XM_017016858.1 | XP_016872347.1 | 4 - 5 | 879 | 112 | |
XM_017016859.1 | XP_016872348.1 | 4 - 5 | 882 | 112 | |
GenBank mRNA | AB000781.1 | - | 3 - 4 | 407 | 112 |
AB051555.1 | - | 3 - 4 | 605 | 112 | |
AK090457.1 | - | 4 - 5 | 474 | 112 |