Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EPHA1
Entrez Gene ID
2041
Gene Name
EPH receptor A1
Gene Aliases
EPH, EPHT, EPHT1
Chromosome Location
Chr.7: 143390813 - 143408892 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005232.4 NP_005223.4 13 - 14 2230 78
XM_006715880.3 XP_006715943.1 12 - 13 1858 78
GenBank mRNA AK290351.1 - 13 - 14 2194 78
AK313263.1 - 13 - 14 2203 78
BC130291.1 - 13 - 14 2214 78
Z27409.1 - 9 - 10 1288 78

View Full Product Details