The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139248.2 | NP_640341.1 | 3 - 4 | 637 | 85 |
XM_006713529.3 | XP_006713592.1 | 3 - 4 | 644 | 85 | |
XM_011512530.2 | XP_011510832.1 | 4 - 5 | 821 | 85 | |
XM_011512531.2 | XP_011510833.1 | 5 - 6 | 766 | 85 | |
GenBank mRNA | AK074229.1 | - | 2 - 3 | 173 | 85 |
AK122651.1 | - | 3 - 4 | 637 | 85 | |
AK298380.1 | - | 3 - 4 | 441 | 85 | |
AY036912.1 | - | 3 - 4 | 617 | 85 | |
AY093498.1 | - | 3 - 4 | 609 | 85 | |
BC064941.1 | - | 3 - 4 | 624 | 85 | |
EF186229.1 | - | 3 - 4 | 614 | 85 |