The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139248.2 | NP_640341.1 | 4 - 5 | 735 | 91 |
XM_011512530.2 | XP_011510832.1 | 5 - 6 | 919 | 91 | |
XM_011512531.2 | XP_011510833.1 | 6 - 7 | 864 | 91 | |
GenBank mRNA | AK074229.1 | - | 3 - 4 | 271 | 91 |
AK122651.1 | - | 4 - 5 | 735 | 91 | |
AY036912.1 | - | 4 - 5 | 715 | 91 | |
AY093498.1 | - | 4 - 5 | 707 | 91 | |
BC064941.1 | - | 4 - 5 | 722 | 91 | |
EF186229.1 | - | 4 - 5 | 712 | 91 |