Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ESRRG
Entrez Gene ID
2104
Gene Name
estrogen related receptor gamma
Gene Aliases
ERR3, ERRgamma, NR3B3
Chromosome Location
Chr.1: 216503246 - 217137757 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134285.2 NP_001127757.1 6 - 7 952 89
NM_001243505.1 NP_001230434.1 4 - 5 392 89
NM_001243506.1 NP_001230435.1 4 - 5 462 89
NM_001243507.1 NP_001230436.1 5 - 6 1239 89
NM_001243509.1 NP_001230438.1 6 - 7 1330 89
NM_001243510.2 NP_001230439.1 6 - 7 946 89
NM_001243511.2 NP_001230440.1 5 - 6 828 89
NM_001243512.1 NP_001230441.1 6 - 7 895 89
NM_001243513.1 NP_001230442.1 5 - 6 915 89
NM_001243514.1 NP_001230443.1 4 - 5 1072 89
NM_001243515.1 NP_001230444.1 6 - 7 1171 89
NM_001243519.1 NP_001230448.1 6 - 7 1175 89
NM_001438.3 NP_001429.2 4 - 5 876 89
NM_206594.2 NP_996317.1 6 - 7 996 89
NM_206595.2 NP_996318.1 5 - 6 878 89
XM_011509266.2 XP_011507568.1 6 - 7 1333 89
XM_017000626.1 XP_016856115.1 6 - 7 1070 89
XM_017000627.1 XP_016856116.1 6 - 7 1133 89
XM_017000638.1 XP_016856127.1 7 - 8 1134 89
XM_017000639.1 XP_016856128.1 6 - 7 974 89
XM_017000640.1 XP_016856129.1 5 - 6 3237 89
XM_017000641.1 XP_016856130.1 5 - 6 1032 89
XM_017000642.1 XP_016856131.1 6 - 7 1130 89
XM_017000643.1 XP_016856132.1 7 - 8 1127 89
XM_017000644.1 XP_016856133.1 6 - 7 1005 89
XM_017000645.1 XP_016856134.1 5 - 6 887 89
XM_017000646.1 XP_016856135.1 6 - 7 3370 89
XM_017000647.1 XP_016856136.1 5 - 6 1119 89
XM_017000648.1 XP_016856137.1 5 - 6 1008 89
XM_017000649.1 XP_016856138.1 7 - 8 1456 89
GenBank mRNA AB020639.1 - 4 - 5 857 89
AB362218.1 - 6 - 7 1249 89
AF058291.1 - 5 - 6 852 89
AF094518.1 - 4 - 5 828 89
AK131193.1 - 5 - 6 1200 89
AK290945.1 - 6 - 7 1152 89
AK291028.1 - 5 - 6 853 89
AK291647.1 - 4 - 5 849 89
AK298851.1 - 4 - 5 429 89
AY388456.1 - 4 - 5 831 89
AY388457.1 - 5 - 6 864 89
AY388458.1 - 6 - 7 890 89
AY388459.1 - 5 - 6 907 89
AY388460.1 - 4 - 5 747 89
AY388461.1 - 4 - 5 1121 89
BC008218.2 - 6 - 7 1035 89
BM723962.1 - 4 - 5 392 89
BX647076.1 - 6 - 7 952 89
CR749497.1 - 6 - 7 945 89
HQ692853.1 - 4 - 5 703 89
HQ692854.1 - 3 - 4 634 89

View Full Product Details