The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170670.1 | NP_001164141.1 | 6 - 7 | 1057 | 79 |
NM_017953.3 | NP_060423.3 | 5 - 6 | 1174 | 79 | |
XM_011541614.2 | XP_011539916.1 | 5 - 6 | 1188 | 79 | |
GenBank mRNA | AF308296.1 | - | 5 - 6 | 821 | 79 |
AK000736.1 | - | 5 - 6 | 546 | 79 | |
AK000767.1 | - | 5 - 6 | 1083 | 79 | |
AK298149.1 | - | 6 - 7 | 1057 | 79 | |
AK314566.1 | - | 5 - 6 | 1102 | 79 | |
AL442074.1 | - | 5 - 6 | 1159 | 79 | |
BC026236.1 | - | 5 - 6 | 1161 | 79 | |
BC110898.1 | - | 5 - 6 | 1092 | 79 |