The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170670.1 | NP_001164141.1 | 8 - 9 | 1202 | 78 |
NM_017953.3 | NP_060423.3 | 7 - 8 | 1319 | 78 | |
XM_011541614.2 | XP_011539916.1 | 7 - 8 | 1333 | 78 | |
GenBank mRNA | AF308296.1 | - | 7 - 8 | 965 | 78 |
AK000736.1 | - | 7 - 8 | 691 | 78 | |
AK000767.1 | - | 7 - 8 | 1228 | 78 | |
AK298149.1 | - | 8 - 9 | 1202 | 78 | |
AK314566.1 | - | 7 - 8 | 1247 | 78 | |
AL442074.1 | - | 7 - 8 | 1304 | 78 | |
BC026236.1 | - | 7 - 8 | 1306 | 78 | |
BC110898.1 | - | 7 - 8 | 1237 | 78 |