The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012128.3 | NP_036260.2 | 9 - 10 | 1506 | 130 |
NR_024602.1 | 8 - 9 | 1397 | 130 | ||
XM_011541015.2 | XP_011539317.1 | 9 - 10 | 1325 | 130 | |
GenBank mRNA | AF127035.1 | - | 9 - 10 | 1490 | 130 |
AK000072.1 | - | 9 - 10 | 1504 | 130 | |
AK000138.1 | - | 2 - 3 | 183 | 130 | |
AK307785.1 | - | 8 - 9 | 1342 | 130 | |
AK307898.1 | - | 7 - 8 | 1111 | 130 | |
AK314430.1 | - | 9 - 10 | 1502 | 130 | |
AY358470.1 | - | 9 - 10 | 1486 | 130 | |
BC113687.1 | - | 9 - 10 | 1501 | 130 | |
BC113689.1 | - | 9 - 10 | 1500 | 130 |