Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LPO
Entrez Gene ID
4025
Gene Name
lactoperoxidase
Gene Aliases
SPO
Chromosome Location
Chr.17: 58238426 - 58268518 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160102.1 NP_001153574.1 5 - 6 843 86
NM_006151.2 NP_006142.1 7 - 8 1092 86
NR_027647.1 6 - 7 1004 86
XM_011524808.2 XP_011523110.1 7 - 8 968 86
GenBank mRNA AK303903.1 - 6 - 7 839 86
AK308378.1 - 7 - 8 817 86
BC107166.1 - 7 - 8 963 86
BC107167.1 - 7 - 8 963 86
BX648309.1 - 7 - 8 930 86
U39573.1 - 7 - 8 857 86

View Full Product Details