Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LPO
Entrez Gene ID
4025
Gene Name
lactoperoxidase
Gene Aliases
SPO
Chromosome Location
Chr.17: 58238426 - 58268518 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160102.1 NP_001153574.1 6 - 7 1174 64
NM_006151.2 NP_006142.1 8 - 9 1423 64
NR_027647.1 7 - 8 1335 64
XM_011524810.2 XP_011523112.1 2 - 3 898 64
GenBank mRNA AK303903.1 - 7 - 8 1170 64
BC107166.1 - 8 - 9 1294 64
BC107167.1 - 8 - 9 1294 64
BX538344.1 - 4 - 5 494 64
BX648309.1 - 8 - 9 1261 64
EF579964.1 - 6 - 7 896 64
U39573.1 - 8 - 9 1188 64

View Full Product Details