The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032824.2 | NP_116213.1 | 13 - 14 | 1637 | 94 |
XM_005263827.2 | XP_005263884.1 | 13 - 14 | 1591 | 94 | |
XM_006712807.2 | XP_006712870.1 | 13 - 14 | 1594 | 94 | |
XM_017005121.1 | XP_016860610.1 | 11 - 12 | 998 | 94 | |
GenBank mRNA | AK027587.1 | - | 13 - 14 | 1622 | 94 |
AK074243.1 | - | 10 - 11 | 895 | 94 | |
AK290294.1 | - | 13 - 14 | 1280 | 94 | |
BC115373.1 | - | 13 - 14 | 1438 | 94 |