The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032824.2 | NP_116213.1 | 14 - 15 | 1742 | 157 |
XM_005263827.2 | XP_005263884.1 | 14 - 15 | 1696 | 157 | |
XM_006712807.2 | XP_006712870.1 | 14 - 15 | 1699 | 157 | |
XM_017005121.1 | XP_016860610.1 | 12 - 13 | 1103 | 157 | |
GenBank mRNA | AK027587.1 | - | 14 - 15 | 1727 | 157 |
AK074243.1 | - | 11 - 12 | 1000 | 157 | |
AK290294.1 | - | 14 - 15 | 1385 | 157 | |
BC115373.1 | - | 14 - 15 | 1543 | 157 |